Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sleeve samplers, U-D Master

Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Rustfrit stål 316L, 1,7 ml
Katalog # SAMP1500W-17
Leverandør:   Sampling Systems
Sleeve samplers, U-D Master
Katalog # SAMP1500W-17
Leverandør:  Sampling Systems
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Rustfrit stål 316L, 1,7 ml

Price
1.300,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The U-D sleeve sampler was developed for accurate unit dose samples (small volume).

  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume

Designed for use with free flowing powders and granules.

Specifikationer
  • Volumen:
    1,7 ml
  • Beskrivelse:
    Tip for U-D sleeve sampler
  • Materiale:
    Rustfrit stål 316L