Jod ≥99.8%, solid ACS, Fluka™
Glass bottle, 1 kg
Leverandør: Honeywell Chemicals
Jod ≥99.8%, solid ACS, Fluka™
Katalog # 207772-1KG
Leverandør: Honeywell Chemicals
CAS nummer: 7553-56-2
Gå til:
Product Details & Documents
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested meet ACS general use specifications
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Smeltende Pt:113,5 °C
- Tæthed:4,930 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-442-4
- FN:3495
- ADR:8,III
Documents
Specifikationer
Specifikationstestresultater
- Assay:Min. 99.8 %
- Non-volatile matter:Max. 0.01 %
- Bromine and chlorine (as chloride):Max. 0.005 %
- Form:Solid
- Vapor Pressure:0.31 mmHg ( 25 °C)
- Resistivity:1.3E15 μΩ-cm
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



