Signs, warning stands
Zone de travail (French), PP, 450×600 mm
Katalog # BRDY256281
Gå til:
Discontinued
Product Details & Documents
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
- Made of high-density polypropylene
- Durable, yellow
- No buckling or corrosion
- Large note field is visible from over 9 meters
- With default notices or unlabeled also available
Height: 660 mm
Specifikationer
- Beskrivelse:Zone de travail (French)
- Dimensioner:450×600 mm
- Materiale:PP
- Seq:07



