Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Detectable scoops, SteriPlast®

Detectable scoops, SteriPlast®

1000 ml, 332 mm, blå
Katalog # 231-0597
Leverandør:   BURKLE
Detectable scoops, SteriPlast®
Katalog # 231-0597
Leverandør:  BURKLE
Detectable scoops, SteriPlast®
Detectable scoops, SteriPlast®

1000 ml, 332 mm, blå

Price
1.420,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

These detectable SteriPlast® scoops, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Detectable scoops are ideal for powders, granulates and pastes.

  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations

These scoops are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.

Specifikationer
  • Kapacitet:
    1000 ml
  • Farve:
    blå
  • Længde:
    332 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.