Cable ties, non-releasable
sort, 3,5×200 mm
BRDY196283
: Brady
Nylon cable ties, easily fixed around items up to 165 mm in circumference.
Ideal for colour-coding hard to mark items.
- Farve:sort
- Dimensioner:3,5×200 mm
- Seq:01