Accessories for various stands
Sedimentation funnel holder for Imhoff funnels with Ø 90 mm or more
Katalog # 612-5699
Leverandør: BURKLE
Gå til:
Product Details & Documents
Funnel holders for Bürkle funnel racks.
Specifikationer
- Beskrivelse:Sedimentation funnel holder for Imhoff funnels with Ø 90 mm or more