Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Blocks for digital heating/cooling block heater

Interchangeable block, 50 ml centrifuge tubes (large), 6, 98×76,5×87 mm
  BARN88880133
Blocks for digital heating/cooling block heater
  BARN88880133
Blocks for digital heating/cooling block heater

Interchangeable block, 50 ml centrifuge tubes (large), 6, 98×76,5×87 mm

 
2.330,00 kr
   
1 * 1 Items
 

Blocks for digital block heater to optimise flexibility and performance.

 
  • Ant. huller:
    6
  • Beskrivelse:
    Interchangeable block
  • Seq:
    04
  • B×D×H:
    98×76,5×87 mm
  • Til:
    50 ml centrifuge tubes (large)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.