Blocks for digital heating/cooling block heater
Interchangeable block, 15 ml centrifuge tubes, 15, 98×76,5×51 mm
Katalog # BARN88880131
Leverandør: Thermo Fisher Scientific
Blocks for digital heating/cooling block heater
Katalog # BARN88880131
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Blocks for digital block heater to optimise flexibility and performance.
Specifikationer
- Ant. huller:15
- Beskrivelse:Interchangeable block
- Seq:02
- B×D×H:98×76,5×51 mm
- Til:15 ml centrifuge tubes
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



