Caps for jars
hvid, HDPE, Jar, 325 ml
Katalog # RIXI8-4120-65
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Slip lids are made of white HDPE.
Specifikationer
- Til:Jar, 325 ml
- Farve:hvid
- Materiale:HDPE