Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Aluminium seals, lined, WHEATON®

Aluminium seals, lined, WHEATON®

Natural, open top, PTFE/grey butyl, 11 mm, aluminium
Catalog # WHEA224235-01
Supplier:   DWK Life Sciences
Aluminium seals, lined, WHEATON®
Catalog # WHEA224235-01
Supplier:  DWK Life Sciences
Aluminium seals, lined, WHEATON®
Aluminium seals, lined, WHEATON®

Natural, open top, PTFE/grey butyl, 11 mm, aluminium

Price
5.120,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Open top seal provides for filling or retrieving contents with a syringe, large target diameter.

  • For use with 12×32 mm crimp top vials
  • Choice of liner

They are also ideal for use as closures in chromatography and instrumentation applications.

Specifications
  • Ø:
    11 mm
  • Septum:
    PTFE/grey butyl
  • Model:
    Natural, open top
  • Materiale:
    aluminium
  • Seq:
    01