Orders LinkContactUsLinkComponent
Labels, energy source, electrical, Brady
Labels, energy source, electrical, Brady
Energy source labels, E-11-19x13mm-SA/100, Polyester, hvid/rød, 19×13 mm
Katalog # BRDY138809
Leverandør:   Brady
Labels, energy source, electrical, Brady
Katalog # BRDY138809
Leverandør:  Brady
Labels, energy source, electrical, Brady
Labels, energy source, electrical, Brady
Energy source labels, E-11-19x13mm-SA/100, Polyester, hvid/rød, 19×13 mm
Price
135,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
These electrical energy source labels are made from laminated polyester.
  • Permanent acrylic adhesive
  • Two different sizes available
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Certifications: ATEX
Emballering: 100 labels per pack or 5 labels per pack.
Specifikationer
  • Farve:
    hvid/rød
  • Beskrivelse:
    Energy source labels
  • Dimensioner:
    19×13 mm
  • Materiale:
    Polyester
  • Seq:
    11
  • Model:
    E-11-19x13mm-SA/100