Group lockout boxes for wall-mounting
Metal group lockout box, small, for 7 padlocks, rød, 203×57,15×178 mm
Katalog # BRDY105714
Leverandør: Brady
Gå til:
Product Details & Documents
These lockout boxes are made of durable, powder-coated steel with key slot on top.
- Clear Lexan window slides out when locks are removed
- Comes with 7 or 15 key pegs that hold 5 keys each and ID labels to simplify key management
- Pre-drilled holes and hardware included for quick mounting
Legend language: English
Specifikationer
- Model:Metal group lockout box, small, for 7 padlocks
- Farve:rød
- Seq:01
- B×D×H:203×57,15×178 mm