Orders LinkContactUsLinkComponent
VWR®, IOS Objectives for Inverted Microscopes
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 4×/0,13
Katalog # 630-2719
Leverandør:   VWR Collection
VWR®, IOS Objectives for Inverted Microscopes
Katalog # 630-2719
Leverandør:  VWR Collection
VWR®, IOS Objectives for Inverted Microscopes
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 4×/0,13
Price
3.700,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These objectives are designed to be used with inverted microscopes.
  • High resolution and contrast
  • Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
  • Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.
Specifikationer
  • Forstørrelse:
    4×/0,13
  • Beskrivelse:
    IOS LWD W-Plan PH objective
  • Seq:
    203