Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux
Fractionating columns, Vigreux
200 mm, 29/32 NS, 29/32 NS
Katalog # WITG1.523.201
Leverandør:   witeg Labortechnik
Fractionating columns, Vigreux
Katalog # WITG1.523.201
Leverandør:  witeg Labortechnik
Fractionating columns, Vigreux
Fractionating columns, Vigreux
200 mm, 29/32 NS, 29/32 NS
Price
950,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Samlet længde:
    200 mm