Accessories
Extended delivery jet assembly, 120 mm, for 1 ml dispenser
Katalog # SOCO1.525.123
Leverandør:   Socorex Isba
Accessories
Katalog # SOCO1.525.123
Leverandør:  Socorex Isba
Accessories
Accessories
Extended delivery jet assembly, 120 mm, for 1 ml dispenser
Price
561,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Extended delivery jet assembly, 120 mm, for 1 ml dispenser
  • Seq:
    17