Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Carbon standards

Organic carbon standard, 20 ppm, 500 mL
Katalog # REGETOC20
Leverandør:   REAGECON
Carbon standards
Katalog # REGETOC20
Leverandør:  REAGECON
CAS nummer:  
Carbon standards

Organic carbon standard, 20 ppm, 500 mL

CAS nummer:  
Price
2.070,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

 : Accredited to IEC/ISO 17025 and ISO 17034.

 

Specifikationer

Specifikationer

  • Koncentration:
    20 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Organic carbon standard

Specifikationer