Orders LinkContactUsLinkComponent
Sampler for Liquids, TeleScoop
Sampler for Liquids, TeleScoop
Sample chamber, Rustfrit stål 316, 100 ml
Katalog # SAMP1035P-100
Leverandør:   Sampling Systems
Sampler for Liquids, TeleScoop
Katalog # SAMP1035P-100
Leverandør:  Sampling Systems
Sampler for Liquids, TeleScoop
Sampler for Liquids, TeleScoop
Sample chamber, Rustfrit stål 316, 100 ml
Price
6.040,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These telescopic handles can be used when sampling from overhead gantries or difficult to reach places.
  • Strong telescopic handle
  • Range of volumes and styles of collecting vessels
Suitable for taking samples from liquids and suspensions.
Specifikationer
  • Kapacitet:
    100 ml
  • Beskrivelse:
    Sample chamber
  • Materiale:
    Rustfrit stål 316