Sampler for Liquids, TeleScoop
Sample chamber, Rustfrit stål 316, 100 ml
Katalog # SAMP1035P-100
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
These telescopic handles can be used when sampling from overhead gantries or difficult to reach places.
- Strong telescopic handle
- Range of volumes and styles of collecting vessels
Suitable for taking samples from liquids and suspensions.
Specifikationer
- Kapacitet:100 ml
- Beskrivelse:Sample chamber
- Materiale:Rustfrit stål 316