VISOCOLOR® Colorimetric Test, Detergents
VISOCOLOR® ECO Detergents anionic, refill pack, Reagents for 50 tests in a box, 0,1 • 0,25 • 0,5 • 1,0 • 2,0 • 5,0 mg/L MBAS, Methylene blue, Visual, 235×130×45 mm (Including packaging), 50, 50 Tests
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Reagents for the determination of detergents in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifikationer
Specifikationer
- Kemisk metode:Methylene blue
- Pakn:50 Tests
- Beskrivelse:VISOCOLOR® ECO Detergents anionic, refill pack
- Dimensioner:235×130×45 mm (Including packaging)
- Til enhed:Visual
- Test pr. sæt:50
- Område:0,1 • 0,25 • 0,5 • 1,0 • 2,0 • 5,0 mg/L MBAS
- Indhold:Reagents for 50 tests in a box
- Cat. No.:MANA931250
- Platform:Colorimetric test kits
- Holdbarhed (måneder):24
- Vægt:415 g (Including packaging)
- Model:MANA931250
- Opbevaringstemperatur:15...25 °C
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



