Orders LinkContactUsLinkComponent
Accessories for cryoscope
Accessories for cryoscope
Sample tubes with 2 ring marks at 2,0 and 2,5 ml
Katalog # GEAG05.5022
Leverandør:   Gerber Instruments AG
Accessories for cryoscope
Katalog # GEAG05.5022
Leverandør:  Gerber Instruments AG
Accessories for cryoscope
Sample tubes with 2 ring marks at 2,0 and 2,5 ml
Price
912,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Sample tubes with 2 ring marks at 2,0 and 2,5 ml
  • Seq:
    06