Forskolin ≥95.0% (by HPLC)
50 mg
Katalog # TCIAF0855-50MG
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS nummer:66575-29-9
- Formel:C₂₂H₃₄O₇
- MDL nummer:MFCD00082317
- Molecular Weight:410.51 g/mol
- Merck Index:14,02476
- EINECS:266-410-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:224.0 to 228.0 deg-C
Specifikationer
- Pakn:50 mg



