Forskolin ≥95.0% (by HPLC)
10 mg
Katalog # TCIAF0855-10MG
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS nummer:66575-29-9
- Formel:C₂₂H₃₄O₇
- MDL nummer:MFCD00082317
- Molecular Weight:410.51 g/mol
- Merck Index:14,02476
- EINECS:266-410-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:224.0 to 228.0 deg-C
Specifikationer
- Pakn:10 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



