Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)

2',5'-Dimethoxypropiophenone ≥98.0% (by GC)

1 g
Catalog # TCIAD4817-1G
Supplier:   TCI
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
Catalog # TCIAD4817-1G
Supplier:  TCI
CAS Number:  5803-30-5
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)

1 g

CAS Number:  5803-30-5
Price
1.090,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(2,5-Dimethoxyphenyl)-1-propanone
  • CAS Number:5803-30-5
  • Formula:C₁₁H₁₄O₃
  • MDL Number:MFCD00027013
  • Molecular Weight:194.23 g/mol
  • Boiling Pt:168°C
  • EINECS:227-357-7
Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specifications

  • Pakn:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.