2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
200 mg
Katalog # TCIAD4817-200MG
Leverandør: TCI
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
Katalog # TCIAD4817-200MG
Leverandør: TCI
CAS nummer: 5803-30-5
Gå til:
Product Details & Documents
Synonymer: 1-(2,5-Dimethoxyphenyl)-1-propanone
- CAS nummer:5803-30-5
- Formel:C₁₁H₁₄O₃
- MDL nummer:MFCD00027013
- Molecular Weight:194.23 g/mol
- Kogende Pt:168°C
- EINECS:227-357-7
Specifikationer
Specifikationstestresultater
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
Specifikationer
- Pakn:200 mg
- Cat. No.:TCIAD4817-200MG
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



