Cover glasses
#1, 8 mm
Katalog # HECH41001108
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
: Available in different diameters.
Specifikationer
- Ø:8 mm
- Antal:#1
- Seq:03