Orders LinkContactUsLinkComponent
Accessories for CryoPlus Sample Storage Systems
Accessories for CryoPlus Sample Storage Systems
Rack includes cardboard boxes and cardboard dividers for 5 ml vials, 676×140×143 mm, for CryoPlus 1, 2, 4
Katalog # THBR4000010
Leverandør:   Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Katalog # THBR4000010
Accessories for CryoPlus Sample Storage Systems
Rack includes cardboard boxes and cardboard dividers for 5 ml vials, 676×140×143 mm, for CryoPlus 1, 2, 4
Price
2.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Rack includes cardboard boxes and cardboard dividers for 5 ml vials, 676×140×143 mm, for CryoPlus 1, 2, 4
  • Seq:
    19