MES monohydrat
Plastic bottle, 1 kg
Leverandør: MERCK PRODUCTION CHEMICALS
Gå til:
Product Details & Documents
2-Morpholinoethanesulfonic acid monohydrate (MES) is suitable for biopharmaceutical production EMPROVE® bio.
Synonymer: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
- CAS nummer:145224-94-8
- Formel:C₆H₁₃NO₄S·1H₂O
- MDL nummer:MFCD00149409
- Molecular Weight:213.25 g/mol
- Smeltende Pt:>300 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:224-632-3
Documents
Specifikationer
Specifikationstestresultater
- Synonyms:MES
- pH value:3,6 (10 g/l, H2O, 20 °C)
- Bulk density:570 kg/m3
- Solubility:80 g/l
- Storage:Store at +5 to +30 °C
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



