Clamps, universal
Clamps, without cork lined jaws, 0 - 80 mm, 150 mm
Katalog # BOCH5425-OK
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These clamps made of stainless steel 18/10 feature cork lined fingers.
Certifications: DIN 12894
Specifikationer
- Spændvidde:0 - 80 mm
- Beskrivelse:Clamps, without cork lined jaws
- Længde:150 mm