Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for drum racks

Accessories for drum racks

Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm, 2 kg
Catalog # CEMG7727
Supplier:   CEMO
Accessories for drum racks
Catalog # CEMG7727
Supplier:  CEMO
Accessories for drum racks
Accessories for drum racks

Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm, 2 kg

Price
350,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Vægt:
    2 kg
  • Beskrivelse:
    Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.