Accessories for drum racks
Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground, 6 kg
Katalog # CEMG7669
Leverandør: CEMO
Gå til:
Product Details & Documents
Specifikationer
- Vægt:6 kg
- Beskrivelse:Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
- Seq:05