Orders LinkContactUsLinkComponent
Accessories for drum racks
Accessories for drum racks
Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums, 1 kg
Katalog # CEMG8254
Leverandør:   CEMO
Accessories for drum racks
Katalog # CEMG8254
Leverandør:  CEMO
Accessories for drum racks
Accessories for drum racks
Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums, 1 kg
Price
384,00 kr
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents
Specifikationer
  • Vægt:
    1 kg
  • Beskrivelse:
    Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums