Orders LinkContactUsLinkComponent
Pallets and sumps
Mobile sump pallet, steel, type SW1, for 1×200 l drum, loading height 490 mm, painted model, loading capacity 300 kg, weight 82,5 kg, 82,5 kg, 980×810×1040 mm
  CEMG7397
 :   CEMO
Pallets and sumps
  CEMG7397
 :  CEMO
Pallets and sumps
Pallets and sumps
Mobile sump pallet, steel, type SW1, for 1×200 l drum, loading height 490 mm, painted model, loading capacity 300 kg, weight 82,5 kg, 82,5 kg, 980×810×1040 mm
7.600,00 kr
1 * 1 Items
The steel sumps provide safe storage for water-polluting as well as inflammable substances.
  • All SW sump pallets are equipped with hot-dip galvanised, removable gratings
  • A wide range of models ensures the correct solution for many application
  • Sump pallets with 100 mm ground clearance permit driving under from all sides
Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.
  • Vægt:
    82,5 kg
  • Beskrivelse:
    Mobile sump pallet, steel, type SW1, for 1×200 l drum, loading height 490 mm, painted model, loading capacity 300 kg, weight 82,5 kg
  • Seq:
    33
  • B×D×H:
    980×810×1040 mm
Accessories
  •  
    Beskrivelse
    Vægt
    B×D×H
  • CEMG7835
    Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 l
    17 kg
    1200×600×400 mm
    1 * 1 Items - 1.630,00 kr
  •  
    Beskrivelse
    Vægt
    B×D×H
  • CEMG7741
    Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 l
    10 kg
    630×450×530 mm
    1 * 1 Items - 1.940,00 kr
  •  
    Beskrivelse
    Vægt
    B×D×H
  • CEMG7586
    Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 l
    15 kg
    600×600×350 mm
    1 * 1 Items - 1.160,00 kr
  •  
    Beskrivelse
    Vægt
    B×D×H
  • CEMG7587
    Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 l
    21 kg
    1130×600×350 mm
    1 * 1 Items - 1.400,00 kr
  •  
    Beskrivelse
    Vægt
    B×D×H
  • CEMG8675
    Drum pallet, PE, for 2×200 l drums
    15 kg
    940×1380×390 mm
    1 * 1 Items - 2.100,00 kr