Orders LinkContactUsLinkComponent
Pallets and sumps
Pallets and sumps
Sump pallet, steel, type SO4, for 1×200 l drum and one small container, painted model, loading capacity 900 kg, weight 56 kg, 1200×1200×265 mm
Katalog # CEMG7681
Leverandør:   CEMO
Pallets and sumps
Katalog # CEMG7681
Leverandør:  CEMO
Pallets and sumps
Pallets and sumps
Sump pallet, steel, type SO4, for 1×200 l drum and one small container, painted model, loading capacity 900 kg, weight 56 kg, 1200×1200×265 mm
Price
2.990,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The steel sumps provide safe storage for water-polluting as well as inflammable substances.
  • All SW sump pallets are equipped with hot-dip galvanised, removable gratings
  • A wide range of models ensures the correct solution for many application
  • Sump pallets with 100 mm ground clearance permit driving under from all sides
Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.
Specifikationer
  • Beskrivelse:
    Sump pallet, steel, type SO4, for 1×200 l drum and one small container, painted model, loading capacity 900 kg, weight 56 kg
  • Seq:
    23
  • B×D×H:
    1200×1200×265 mm
Accessories
Produktinformation
  • Katalog #
    Vægt
    B×D×H
    Tilgængelighed
    Price
  • Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 l
    Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 l
    CEMG7835
    17 kg
    1200×600×400 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.630,00 kr
Produktinformation
  • Katalog #
    Vægt
    B×D×H
    Tilgængelighed
    Price
  • Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 l
    Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 l
    CEMG7741
    10 kg
    630×450×530 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.940,00 kr
Produktinformation
  • Katalog #
    Vægt
    B×D×H
    Tilgængelighed
    Price
  • Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 l
    Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 l
    CEMG7586
    15 kg
    600×600×350 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.160,00 kr
Produktinformation
  • Katalog #
    Vægt
    B×D×H
    Tilgængelighed
    Price
  • Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 l
    Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 l
    CEMG7587
    21 kg
    1130×600×350 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.400,00 kr
Produktinformation
  • Katalog #
    Vægt
    B×D×H
    Tilgængelighed
    Price
  • Drum pallet, PE, for 2×200 l drums
    Drum pallet, PE, for 2×200 l drums
    CEMG8675
    15 kg
    940×1380×390 mm
    Log ind for tilgængelighed
    1 * 1 Items - 2.100,00 kr