Pallets and sumps, PE
Sump pallet 240/4, for 4×242 l drums, with PE grating, load capacity 1000 kg, 31 kg, 1380×1290×280 mm
CEMG8524
: CEMO
These sumps are manufactured from high quality polyethylene (HD-LMD).
- With general construction inspection approval no. Z-40.22-380
- High chemical resistance
Sump pallets 240 have removable perforated plate and the forklift-accessible on all four sides is possible only in 240/4.
Sump pallet 230/2 are suitable for Euro-pallets (1200×800 mm) and plastic pallets of similar size. These are loaded pallets may be placed by a forklift directly on the tray (PE perforated plate is not required).
- Vægt:31 kg
- Beskrivelse:Sump pallet 240/4, for 4×242 l drums, with PE grating, load capacity 1000 kg
- Seq:04
- B×D×H:1380×1290×280 mm
Accessories
- BeskrivelseVægtB×D×H
- CEMG10519Access ramp for drum depot PE-250-4-GH, load capacity 500 kg27 kg1750×750×300 mm1 * 1 Items - 3.140,00 kr
- BeskrivelseVægtB×D×H
- CEMG10518Access ramp, for drum depot PE 250-2-GH, load capacity 500 kg54 kg2500×750×450 mm1 * 1 Items - 5.920,00 kr
- BeskrivelseVægtB×D×H



