Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Pallets and sumps, PE

Sump pallet 240/1, for 1×225 l drum, with PE grating, load capacity 250 kg, 17 kg, 1000×680×600 mm
  CEMG10017
 :   CEMO
Pallets and sumps, PE
  CEMG10017
 :  CEMO
Pallets and sumps, PE
Pallets and sumps, PE

Sump pallet 240/1, for 1×225 l drum, with PE grating, load capacity 250 kg, 17 kg, 1000×680×600 mm

 
 
   
1 * 1 Items
 

These sumps are manufactured from high quality polyethylene (HD-LMD).

  • With general construction inspection approval no. Z-40.22-380
  • High chemical resistance

Sump pallets 240 have removable perforated plate and the forklift-accessible on all four sides is possible only in 240/4.

Sump pallet 230/2 are suitable for Euro-pallets (1200×800 mm) and plastic pallets of similar size. These are loaded pallets may be placed by a forklift directly on the tray (PE perforated plate is not required).

 
  • Vægt:
    17 kg
  • Beskrivelse:
    Sump pallet 240/1, for 1×225 l drum, with PE grating, load capacity 250 kg
  • Seq:
    01
  • B×D×H:
    1000×680×600 mm
Accessories

 

  •  
    Beskrivelse
    Vægt
    B×D×H
     
     
  • CEMG10519
    Access ramp for drum depot PE-250-4-GH, load capacity 500 kg
    27 kg
    1750×750×300 mm
       
    1 * 1 Items - 3.140,00 kr

 

  •  
    Beskrivelse
    Vægt
    B×D×H
     
     
  • CEMG10518
    Access ramp, for drum depot PE 250-2-GH, load capacity 500 kg
    54 kg
    2500×750×450 mm
       
    1 * 1 Items - 5.920,00 kr

 

  •  
    Beskrivelse
    Vægt
    B×D×H
     
     
  • CEMG10286
    Perforated plate for PE sump pallet for type 230/2
    8 kg
    1215×815×45 mm
       
    1 * 1 Items

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.