Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Erlenmeyer Flasks, with Screw Cap

VWR®, Erlenmeyer Flasks, with Screw Cap

150 ml, 74 mm, GL 25
Katalog # 214-0409
Leverandør:   VWR Collection
VWR®, Erlenmeyer Flasks, with Screw Cap
Katalog # 214-0409
Leverandør:  VWR Collection
VWR®, Erlenmeyer Flasks, with Screw Cap
VWR®, Erlenmeyer Flasks, with Screw Cap

150 ml, 74 mm, GL 25

Price
193,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Borosilicate 3.3 glass, blue PP screw cap with PTFE liner.

  • Highly resistant to chemicals and thermal shock
  • Graduations and white labelling area
  • Delivered with blue PP screw cap

Ideal for storage or transportation of dangerous liquids. They are leakproof and can be autoclaved together with the screw cap at 140 °C.

Be aware that the screw cap should not be closed during the autoclaving process.

Specifikationer
  • Udv.Ø:
    74 mm
  • Gevind:
    GL 25
  • Kapacitet:
    150 ml
  • Seq:
    03