Microplate washers, AquaMax®
Product Details & Documents
AquaMax® microplate washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
- Wash heads can be changed in 30 seconds with no tools using innovative head latch mechanism
- All replacement heads are recognised by the washer base, no recalibration required
- Simultaneous well washing without plate indexing or quadrant processing
- Completely programmable via touch screen interface
- No external pumps or computer
- Comprehensive, automated cleaning utilities
- Robotics-friendly design
AquaMax® washers have several important automatic maintenance protocols, since the lack of proper washer maintenance is the number one cause of washer failure. One-Touch AutoClean completely washes system with AquaMax® sterilant; rinses with water and then air dries both the aspiration and dispensing channel to prevent contaminant build up. Auto-shutdown is user-definable and can automatically rinse and purge the AquaMax® washer with air, providing walk away flexibility at the end of the batch.
Delivery information: AquaMax® washer is available in two configurations. The AquaMax® 2000 has two fluid inlets; the AquaMax® 4000 has four fluid inlets and additional programming capabilities. Both systems have the same onboard touch screen computer for all programming, operation, and maintenance of the system. No external computer is necessary.
Documents
Specifications
- Beskrivelse:AquaMax® 2K includes AquaMax® 2000 microplate washer, 96-plate wash head, 2 assay bottles with level sensor, and 10 l waste bottle with sensor
- Cat. No.:735-0382
- Dispenseringsvolumenområde:10 - 400 µl
- Model:AquaMax®
- Pladetyper:96- and 384-well
- Interface:USB and proprietary StakMax connect
Complementary Products
Product Information
- Catalog #BeskrivelseAnt. brøndeSterilFarveBrøndvolumenOpløsningModelAvailabilityPrice
- MLDVPARADIGMSpectraMax® Paradigm®, microplate reader, multi-modeLog in for availability1 * 1 Items - 0,00 kr
- 735-0358SpectraMax® iD5 multi-mode microplate reader, with 20×20 well scanningLog in for availability1 * 1 Items - 0,00 kr
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



