Orders LinkContactUsLinkComponent
Cell integrity kit, EarlyTox™
Cell integrity kit, EarlyTox™
EarlyTox™ Live Red Dye, 1 vial, sufficient for two 96- or 384- well plates, 1 KIT
Katalog # MLDVR8217
Leverandør:   Molecular Devices
Cell integrity kit, EarlyTox™
Katalog # MLDVR8217
Leverandør:  Molecular Devices
CAS nummer:  
Cell integrity kit, EarlyTox™
Cell integrity kit, EarlyTox™
EarlyTox™ Live Red Dye, 1 vial, sufficient for two 96- or 384- well plates, 1 KIT
CAS nummer:  
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The EarlyTox™ Cell Integrity Kit enables the differentiation of live cells from dead cells via fluorescent labeling. This is useful for the rapid quantification of cell viability when used with the SpectraMax® MiniMax™ 300 Imaging Cytometer or other fluorescence-based cellular imaging instruments.
  • Useful for the rapid quantification of cell viability
  • Higher signal allows shorter exposure times for fast results
  • Designed to work with many cell types - both adherent and non adherent
  • Streamlined image acquisition and analysis on a single system
The EarlyTox® Cell Integrity Kit is an optimised set of reagents that simplifies the identification of live and dead cells. It can be used to measure the effects of different treatments on cell viability and to evaluate toxic effects mediated through a variety of mechanisms, including apoptosis and necrosis. The kit is designed to work with many cell types, both adherent and non-adherent. The protocol's simple workflow and reagent performance make it amenable to high-throughput screening.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    EarlyTox™ Live Red Dye, 1 vial, sufficient for two 96- or 384- well plates