Bootie benches
Bootie bench with 2 solid undershelves and dividers, 1400×473×550 mm
Katalog # PALBBB1447-2D
Leverandør: Palbam
Gå til:
Product Details & Documents
Bootie benches are designed as a swing over bench and a storage solution for booties/shoes.
- Made of stainless steel 304L with electro-polished finish
- Available in three options according to the length required
Documents
Palbam Class - Catalog 2018
Produktlitteratur
Specifikationer
- Beskrivelse:Bootie bench with 2 solid undershelves and dividers
- B×D×H:1400×473×550 mm