Orders LinkContactUsLinkComponent
Detectable scoops, SteriPlast®
500 ml, 280 mm, blå
  231-0578
 :   BURKLE
Detectable scoops, SteriPlast®
  231-0578
 :  BURKLE
Detectable scoops, SteriPlast®
Detectable scoops, SteriPlast®
500 ml, 280 mm, blå
919,00 kr
1 * 10 Items
These detectable SteriPlast® scoops, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Detectable scoops are ideal for powders, granulates and pastes.
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
These scoops are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
  • Kapacitet:
    500 ml
  • Farve:
    blå
  • Længde:
    280 mm
  • Seq:
    06