Detectable scoops, SteriPlast®
Product Details & Documents
These detectable SteriPlast® scoops, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Detectable scoops are ideal for powders, granulates and pastes.
- Can be detected by colour scanners, metal detectors or X-rays
- Individually packaged and sterilised by gamma rays
- Production, assembly and packaging according to cleanroom class 7
- Complies with EU food requirements and FDA regulations
These scoops are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Specifikationer
- Kapacitet:100 ml
- Farve:blå
- Længde:205 mm
- Seq:03
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



