Dextran, M.W. ~ 70000 (Dextran 70)
100 g
Katalog # TCIAD1449-100G
Leverandør: TCI
Gå til:
Product Details & Documents
- CAS nummer:9004-54-0
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00130935
- Merck Index:14,02948
- EINECS:232-677-5
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to Almost white powder to crystaline
- Specific rotation [a]20/D:+170 to +200 deg (C=6, H2O)
- pH:5.0 to 7.0 (60g/L, 25deg-C)
- limiting viscosity:0.21 to 0.26 cp
Specifikationer
- Pakn:100 g



