ARISTAR®, Bismuth Standard Solutions for ICP-MS
Vismut, Bi, 10 ppm, 2-5% HNO₃, Plastic bottle, 100 mL
Leverandør: VWR Chemicals
ARISTAR®, Bismuth Standard Solutions for ICP-MS
Katalog # 85554.180
Leverandør: VWR Chemicals
CAS nummer:
Gå til:
Product Details & Documents
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Documents
Certifikater
Søg efter partinummer125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125)
Emballageoplysninger
VWR® for Calibrations
Produktlitteratur
Specifikationer
Specifikationer
- Koncentration:10 ppm
- Matrice:2-5% HNO₃
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Vismut
- Symbol:Bi
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



