Orders LinkContactUsLinkComponent
Fluid samplers, Liquid Master
Fluid samplers, Liquid Master
Fluid sampler, Rustfrit stål 316, 420 ml, 1500 mm, 25 mm
Catalog # SAMP5035S-1500
Supplier:   Sampling Systems
Fluid samplers, Liquid Master
Catalog # SAMP5035S-1500
Supplier:  Sampling Systems
Fluid samplers, Liquid Master
Fluid samplers, Liquid Master
Fluid sampler, Rustfrit stål 316, 420 ml, 1500 mm, 25 mm
Price
9.140,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The Liquid Master has been designed to be quickly stripped down for cleaning.
  • Quick and easy to use
  • Push button to press to take samples
  • For a larger volume, the sampler can be inserted deep into the product
Suitable for taking samples from low and medium viscosity liquids.
Specifications
  • Ø:
    25 mm
  • Kapacitet:
    420 ml
  • Beskrivelse:
    Fluid sampler
  • Længde:
    1500 mm
  • Materiale:
    Rustfrit stål 316
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0492
    Nylon
    15 - 30 mm
    1100 mm
    Log in for availability
    1 * 1 Items - 418,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0514
    Nylon
    7 - 14 mm
    1600 mm
    Log in for availability
    1 * 1 Items - 501,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0493
    Nylon
    15 - 30 mm
    1600 mm
    Log in for availability
    1 * 1 Items - 501,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0494
    Nylon
    15 - 30 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0513
    Nylon
    7 - 14 mm
    1100 mm
    Log in for availability
    1 * 1 Items - 418,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0495
    Nylon
    15 - 30 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0496
    Nylon
    30 - 45 mm
    1100 mm
    Log in for availability
    1 * 1 Items - 418,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0497
    Nylon
    30 - 45 mm
    1600 mm
    Log in for availability
    1 * 1 Items - 501,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0498
    Nylon
    30 - 45 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0499
    Nylon
    30 - 45 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0516
    Nylon
    7 - 14 mm
    600 mm
    Log in for availability
    1 * 1 Items - 334,00 kr
Product Information
  • Catalog #
    Børstemateriale
    Børstehoved Ø
    Samlet længde
    Availability
    Price
  • 300-0515
    Nylon
    7 - 14 mm
    2100 mm
    Log in for availability
    1 * 1 Items - 585,00 kr