Silikoneslanger
Size 73, with hose barb of 9,5 mm, 9,50 mm, 73
SPECACTU-P73-25N
Silicone, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Rør int.Ø:9,50 mm
- Beskrivelse:Size 73, with hose barb of 9,5 mm
- Seq:10
- Størrelse:73