Temperature Probes for Heating Mantle Controller
Temperature probes SS310, PTFE-coated, 4,8×250 mm
Katalog # WITGDH.WHP503041
Leverandør: witeg Labortechnik
Temperature Probes for Heating Mantle Controller
Katalog # WITGDH.WHP503041
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Temperature probe for use with heating mantle controller WHM-C10D.
Specifikationer
- Beskrivelse:Temperature probes SS310, PTFE-coated
- Seq:3
- Ø×L:4,8×250 mm