Orders LinkContactUsLinkComponent
Temperature Probes for Heating Mantle Controller
Temperature Probes for Heating Mantle Controller
Temperature probes SS310, PTFE-coated, 4,8×250 mm
Katalog # WITGDH.WHP503041
Leverandør:   witeg Labortechnik
Temperature Probes for Heating Mantle Controller
Katalog # WITGDH.WHP503041
Leverandør:  witeg Labortechnik
Temperature Probes for Heating Mantle Controller
Temperature probes SS310, PTFE-coated, 4,8×250 mm
Price
1.730,00 kr
Log ind for tilgængelighed
1 * 1 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Temperature probe for use with heating mantle controller WHM-C10D.
Specifikationer
  • Beskrivelse:
    Temperature probes SS310, PTFE-coated
  • Seq:
    3
  • Ø×L:
    4,8×250 mm