Ion selective meter, handheld, Eutech Ion 6+
Eutech Ion 6+ Ion/pH/mV meter with single junction pH electrode ECFC7252101B, ATC electrode ECPHTEM01P and pH carrying kit (incl. 1× pH 4,01 + pH 7,00 buffer solutions, storage solution and 1× deionised (rinse) water (60 ml each))
Katalog # 662-2288
Leverandør: EUTECH
Gå til:
Product Details & Documents
The portable Eutech Ion 6+ offers measurements for pH, ORP, temperature and ion concentration. With its compact and sturdy design, the lightweight meter is the ideal choice for field and lab work.
- Convenient and easy to use with pre-programmed Ion calibration points
- mV mode allows users to check electrode calibration and performance
- Offers a wide measurement range of up to ±500 mV
- Compatible with a large variety of ISE electrodes
- Shock-resistant ABS plastic casing, protected by an IP54-rated housing against accidental splashes
- HOLD function, automatic power-off after 20 minutes of inactivity
- Convenient benchtop stand, tactile keypad and customised LCD screen
Typical applications include: Pools and spas, ponds and aquariums, agriculture and hydroponics, water and wastewater treatment, cooling towers, food processing, photo-development and printing, ecological studies, field, laboratory and education institutions.
Documents
Eutech 5+ & 6+ Series Economy Handheld
Produktlitteratur
Specifikationer
- Beskrivelse:Eutech Ion 6+ Ion/pH/mV meter with single junction pH electrode ECFC7252101B, ATC electrode ECPHTEM01P and pH carrying kit (incl. 1× pH 4,01 + pH 7,00 buffer solutions, storage solution and 1× deionised (rinse) water (60 ml each))
- Seq:02
- mV - Område:±500
- pH - Aflæsningsinterval:0,01
- Strømforsyning:4× 1,5 V ‘AAA’ alkaline batteries
- Overensstemmelse:IP54
- pH - Nøjagtighed:±0,01
- Temperaturinterval:0,0...+100,0 °C
- ISE Område:0,01 - 0,99 ppm / 1,0 - 199,9 ppm / 200 - 1999 ppm
- pH - Område:0,00 - 14,00
- pH - Kalibrering:Up to 3 points
- Temperature resolution:0,1 °C
- Tilslutninger:BNC, 2,5 mm phono socket
- ISE Kalibrering:Up to 3 points
- Cat. No.:662-2288
- ISE Nøjagtighed:±1 % full scale
- ISE Opløsning:0,01 / 0,1 / 1 ppm
- Temperaturkompensation:ATC/MTC (0...+100 °C)
- Vægt:0,26 kg
- Model:Eutech Ion 6+
- W×D×H:85×42×157 mm
- mV - Aflæsningsinterval:0,1 (±199,9); 1 (beyond)
- mV - Nøjagtighed:±0,2 (±199,9); ±2 (beyond)
- Display:Customised LCD
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



