Accessories for microcarriers
Aseptic transfer cap, PP, orange, disposable ,48 mm, MPC, for 500 g microcarrier bottle
Katalog # 734-2922
Leverandør: Corning
Gå til:
Product Details & Documents
These polypropylene aseptic transfer screw caps prevents adventitious contamination in for microcarriers bottles during bioreactor loading.
- Chemically resistant and heat sealable flexible tubing
- Tubing length with connector 30" (76,2 cm)
- Disposable and sterile (10⁻⁶ SAL)
Emballering: These caps are individually double bagged.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Beskrivelse:Aseptic transfer cap, PP, orange, disposable ,48 mm, MPC, for 500 g microcarrier bottle
- Cat. No.:734-2922
- Miljøcertificering:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Lav fremstillingspåvirkning