Accessories for CryoPlus Sample Storage Systems
Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
Katalog # THBR4000042
Leverandør: Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Katalog # THBR4000042
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
- Seq:15