Orders LinkContactUsLinkComponent
Cryoboxes, PP
Cryoboxes, PP
7×7, blå, 133×133×50/75 mm
Katalog # RATI5320314
Leverandør:   RATIOLAB
Cryoboxes, PP
Katalog # RATI5320314
Leverandør:  RATIOLAB
Cryoboxes, PP
Cryoboxes, PP
7×7, blå, 133×133×50/75 mm
Price
290,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Boxes made of high quality PP.
  • Autoclavable
  • Temperature-resistant from –90 to +121 °C
  • Numerically coded
  • Very good chemical resistance
With an numerical coding and corresponding vents for draining condensed water. The boxes are adjustable in height.
Specifikationer
  • Rum:
    7×7
  • B×D×H udv.:
    133×133×50/75 mm
  • Farve:
    blå
  • Seq:
    02