AlphaTec® MICROGARD 2000 Isolation Gowns
hvid, S
Katalog # ANSEWH20B-00214-02
Leverandør: Ansell
Gå til:
Product Details & Documents
Superior breathable microporous laminate technology.
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Silicone-free
- Low-linting
The MICROGARD 2000 range is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0,01 µm in size. Bound seams offer superior strength, as well as a liquid and particle barrier.
These isolation gowns feature rear Velcro fastening, elasticated wrists, and bound seams.
Specifikationer
- Tøj størrelse:S
- Farve:hvid
- Seq:01
- Cat. No.:ANSEWH20B-00214-02
- Sleeve or leg length:Long
- Model:Microgard 2000
- Outfit applikationer:Medical|Clinical
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



